Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Phospho-Tyrosine, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB11015088 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11015088 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11015088 Supplier Novus Biologicals Supplier No. NB11015088

Rabbit Polyclonal Antibody

Phospho-Tyrosine Polyclonal specifically detects Phospho-Tyrosine in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen Phospho-Tyrosine
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, ELISA 1:5000-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 2-5 ug, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS (pH 7.4) with 0.09% Sodium Azide
Gene Alias 2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, phosphotyrosine, pTyrosine, Tyrosine
Host Species Rabbit
Immunogen Affinity purified native tyrosine phosphorylated proteins
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Minimal to no crossreactivity with phosphothreonine or phosphoserine containing proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.