Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-tyrosine Mouse anti-All, Biotin, Clone: 4G10™, MilliporeSigma™

Catalog No. 16204MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-204-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-204-MI Supplier MilliporeSigma Supplier No. 16204
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Phosphotyrosine using this Anti-Phosphotyrosine Antibody, recombinant clone 4G10™, biotin conjugate validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphotyrosine
Applications Western Blot
Classification Monoclonal
Clone 4G10™
Conjugate Biotin
Formulation Biotin-conjugated recombinant mouse IgG2bκ in PBS containing 0.05% sodium azide. Liquid at 2-8°C.
Host Species Mouse
Immunogen Phosphotyramine-KLH
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species All species
Content And Storage 2-8°C for 9 months. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.