Learn More
PKA (RIIα) Mouse, Unlabeled, Clone: 40, BD
Mouse Monoclonal Antibody
$281.50 - $556.00
Specifications
Antigen | PKA (RIIα) |
---|---|
Clone | 40 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
cAMP-dependent Protein Kinase (PKA) is composed of two distinct subunits: catalytic(C) and regulatory (R). Four regulatory subunits have been identified: RIα, RIβ, RIIα, and RIIβ.These subunits define type I and II cAMP-dependent protein kinases. Following binding of cAMP, the regulatory subunits dissociate from the catalytic subunits, rendering the enzyme active. Type I and type II holoenzymes have three potential C subunits (Cα, Cβ, or Cγ). Type II PKA can be distinguished by autophosphorylation of the R-subunits, while type I PKA binds Mg/ATP with high affinity. Most cells express both type I and type II PKAs. Although the Rα isoforms are ubiquitously expressed, the Rβ isoforms are predominant in nervous and adipose tissues. Expression of the RIα subunit is modulated during muscle and adipocyte differentiation in vitro.
Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
PKA (RIIα) | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
40 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human, Mouse, Rat | |
Human PKA[RIIα] aa. 1-404 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.