Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PACT Rabbit anti-Human, Polyclonal, Invitrogen™

Catalog No. PIPA514832
Change view
Click to view available options
Quantity:
400 μL
Catalog No. Quantity
PIPA514832 400 μL
1 options

Catalog No. PIPA514832

Supplier: Thermo Scientific PA514832

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine and rat based on sequence homology.

PRKRA appears to have a pro-apoptotic function that may be suppressed in the presence of growth factor. It activates EIF2AK2 in the absence of double-stranded RNA (dsRNA).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PACT
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene PRKRA
Gene Accession No. O75569
Gene Alias Interferon-inducible double stranded RNA-dependent protein kinase activator A; Protein kinase, interferon-inducible double stranded RNA-dependent activator; Protein activator of the interferon-induced protein kinase; PKR-associated protein X; PKR-associating protein X; PRKRA; PACT; RAX
Gene Symbols PRKRA
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 225-252 amino acids from human PRKRA
Purification Method Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8575
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.