Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTP1C/SHP1 Mouse, Unlabeled, Clone: 52, BD

Catalog No. BDB610126 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610126 150 μg
BDB610125 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610126 Supplier BD Biosciences Supplier No. 610126
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PTP1C (SH-PTP1, SHP-1, or HCP) is a protein-tyrosine phosphatase (PTP) that is expressed in epithelial and hematopoietic cells. It contains two N-terminal SH2 domains and a C-terminal phosphatase domain. While its distribution is primarily cytosolic, exposure to various stimuli induces movement of PTP1C to other cellular locations to allow interaction with its substrates. This enzyme associates with the IL-3, IL-4, erythropoietin, and stem cell factor receptors and may regulate their signaling pathways by dephosphorylation of their receptors or their downstream effectors. Motheaten mice, which possess a nonfunctional PTP1C gene, display multiple hematopoietic abnormalities including increased proliferation and activation of myeloid cells. Studies involving these mice have clearly implicated PTP1C in negative regulation of CSF-1 (colony stimulating factor-1) mitogenic signaling and have demonstrated a role for PTP1C as an intermediate between Ras and the MAP kinase pathway.

Host Species: Mouse
Clone: 52
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human PTP1C aa.. 492-597
Formula Weight [Chemical]: 68kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen PTP1C/SHP1
Applications Western Blot
Classification Monoclonal
Clone 52
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human PTP1C aa.. 492-597
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.