Learn More
Rab11 Mouse, Unlabeled, Clone: 47, BD
Mouse Monoclonal Antibody
$287.50 - $568.50
Specifications
Antigen | Rab11 |
---|---|
Clone | 47 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
The Rab proteins are small GTP-binding molecules that are localized to specific intracellular vesicles and organelles. It has been proposed that Rab proteins cycle between GTP- and GDP-bound forms and that this is related to their function as regulators of vesicular traffic. The Rab11 gene encodes a 24 kDa protein of 214 amino acids that has been detected in liver, brain, testis, spleen, and heart. Rab11 protein was isolated from the golgi-microsomal fraction of rat liver and has been detected in the Trans-golgi Network, secretory vesicles, and the pericentriolar recycling endosomes. The distribution of Rab11 indicates that this small protein is involved in regulating traffic at the Golgi complex.
Specifications
Rab11 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2a | |
Affinity Purified |
47 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Chicken, Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.