Learn More
anti-Rabbit IgG, biotinylated speciesspecific whole antibody (from donkey), Cytiva
Shop All Cytiva ProductsDescription
Host Species: Donkey
Isotype: IgG
Species Reactivity: Rb
Protein Blotting, ELISA, Immunocytochemistry
Specifications
Specifications
| Antigen | IgG, Whole Molecule |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Host Species | Donkey |
| Purification Method | Affinity Purified |
| Quantity | 2 mL |
| Primary or Secondary | Primary |
| Isotype | IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.