Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TTF1 / NKX2.1, Clone: EP1584Y, Novus Biologicals™
SDP

Catalog No. NB10080062 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10080062 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10080062 Supplier Novus Biologicals Supplier No. NB10080062

Specifications

Antigen TTF1 / NKX2.1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone EP1584Y
Conjugate Unconjugated
Gene Accession No. P43699
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues near the N-terminus of human TTF1 was used as an immunogen.
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7080
Target Species Human, Mouse, Rat
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.