Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rabies Virus Monoclonal Antibody (1C5)
GSA_VAGREENER_CHOICE

Catalog No. PIMA121549
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA121549 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA121549 Supplier Invitrogen™ Supplier No. MA121549
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Rabies virus (Neurotropic lyssavirus) is a member of the Rhabdoviridae family. Rabies is a single stranded, neurotropic, negative sense RNA virus which encodes 5 proteins: a glycoprotein, a nucleoprotein, and three others. The mature virus has a bullet shape, a protein coat, and a lipid envelope. The outer surface of the virus is covered with thumblike glycoprotein projections 5-10 nm long and 3 nm in diameter. The virus averages approximately 780 nm in length. Virus infectivity is destroyed by lipid solvents. Rabies virus is a very successful virus, with a very wide range of hosts. It causes an acute, central nervous system infection, characterized by CNS irritation, followed by paralysis and death. Approximately 50,000 human deaths each year are caused by rabies.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabies Virus
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 1C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias Neurotropic lyssavirus
Host Species Mouse
Immunogen Russian rabies vaccine strain Vnukovo-32.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.