Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD19, Biotin, Clone: 6D5, Novus Biologicals™
SDP

Catalog No. NBP127893 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP127893 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP127893 Supplier Novus Biologicals Supplier No. NBP127893

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Antigen CD19
Applications Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), In vitro Assay
Classification Monoclonal
Clone 6D5
Concentration 0.5mg/mL
Conjugate Biotin
Dilution Flow Cytometry <= 1 ug/10^6 cells, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay
Gene Alias B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Gene Symbols CD19
Host Species Rat
Immunogen Mouse CD19-expressing K562 human erythroleukemia cells
Purification Method Protein A or G purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 930
Test Specificity CD19 B-cell differentiation antigen, Mr 95 kDa
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.