Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DcTRAILR2/TNFRSF22 Rat anti-Mouse, Clone: Lucy-1, Novus Biologicals™
SDP

Catalog No. NBP197591 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197591 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197591 Supplier Novus Biologicals Supplier No. NBP197591

Rat Monoclonal Antibody

DcTRAILR2/TNFRSF22 Monoclonal specifically detects DcTRAILR2/TNFRSF22 in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation (Negative).

Specifications

Antigen DcTRAILR2/TNFRSF22
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone Lucy-1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:1000, Immunoprecipitation (Negative)
Gene Symbols TNFRSF25
Host Species Rat
Immunogen Recombinant mouse DcTRAIL-R2 extracelluar domain (aa 1-170).
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8718
Test Specificity Recognizes mouse DcTRAIL-R2. Does not cross-react with mouse DcTRAIL-R1.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.