Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MAdCAM-1, FITC, Clone: MECA-367, Novus Biologicals™
SDP

Catalog No. NBP128147 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128147 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128147 Supplier Novus Biologicals Supplier No. NBP128147

Rat Monoclonal Antibody

MAdCAM-1 Monoclonal specifically detects MAdCAM-1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen MAdCAM-1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MECA-367
Concentration 0.5mg/mL
Conjugate FITC
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500
Gene Alias hMAdCAM-1, MACAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1
Gene Symbols MADCAM1
Host Species Rat
Immunogen Endothelial cells isolated from BALB/c mesenteric and peripheral lymph nodes
Purification Method Affinity Purified
Quantity 0.25 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8174
Test Specificity Murine MAdCAM-1 (Mr 58-88 kDa)
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.