Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Respiratory Syncytial Virus, Clone: 130-12H, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB8583F/DEL
Description
ELISA, Immunofluorescence

Specifications
Respiratory Syncytial Virus | |
FITC | |
Respiratory Syncytial virus A2 | |
RUO | |
Nuclear Protein, 42-44kD | |
Human | |
IgG2a κ |
Monoclonal | |
Mouse | |
100 μg | |
Viral And Bacterial | |
Primary | |
Semi-purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction