Learn More
MilliporeSigma™ anti-Respiratory Syncytial Virus, Clone: 130-12H, ™ (Chemicon™)
Specifications
Antigen | Respiratory Syncytial Virus |
---|---|
Classification | Monoclonal |
Form | Semi-purified |
Host Species | Mouse |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
50-175-803
|
MilliporeSigma™
MAB8583F/DEL |
FITC | 100 μg |
Item Discontinued
This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. View Alternate Product |
N/A | ||||
50-175-800
|
MilliporeSigma™
MAB8583/DEL |
Unconjugated | 100 μg |
Item Discontinued
This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. View Alternate Product |
N/A | ||||
Description
ELISA, Immunofluorescence
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Respiratory Syncytial Virus | |
Semi-purified | |
RUO | |
Human | |
Nuclear Protein, 42-44kD |
Monoclonal | |
Mouse | |
Viral And Bacterial | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.