Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-RPB2, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP220207 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220207 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220207 Supplier Novus Biologicals Supplier No. NBP220207

Rabbit Polyclonal Antibody

RPB2 Polyclonal specifically detects RPB2 in Human samples. It is validated for Western Blot.

Specifications

Antigen RPB2
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P30876
Gene Alias DNA-directed RNA polymerase II subunit RPB2, EC 2.7.7, EC 2.7.7.6, hRPB140, hsRPB2, POL2RB, polymerase (RNA) II (DNA directed) polypeptide B (140kD), polymerase (RNA) II (DNA directed) polypeptide B, 140kDa, RNA polymerase II subunit 2, RNA polymerase II subunit B2, RNA polymerase II subunit2, RNA polymerase II second largest subunit
Gene Symbols POLR2B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human RPB2. The exact sequence is proprietary.
Molecular Weight of Antigen 134 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5431
Test Specificity Human. Species reactivity based on sequence identity: Mouse (100%), Chicken (100%), Rat (100%), Bovine (100%), Rhesus Monkey (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.