Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Salmonella, Biotin, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB10064498 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064498 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064498 Supplier Novus Biologicals Supplier No. NB10064498

Specifications

Antigen Salmonella
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 4.0mg/mL
Conjugate Biotin
Dilution Immunocytochemistry/Immunofluorescence 1:10-1:2000
Formulation PBS with 0.05% Sodium Azide
Host Species Rabbit
Immunogen Made to Bacterial SALMONELLA GROUP ANTIGEN
Purification Method Immunogen affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity SALMONELLA
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.