Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selenoprotein W Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP149599 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149599 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149599 Supplier Novus Biologicals Supplier No. NBP149599

Rabbit Polyclonal Antibody

Selenoprotein W Polyclonal specifically detects Selenoprotein W in Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen Selenoprotein W
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, ELISA 1:100000, Immunoprecipitation 1:100
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Alias selenoprotein W, selenoprotein W, 1, SELW
Gene Symbols SEPW1
Host Species Rabbit
Immunogen This affinity purified Selenoprotein W Antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant protein corresponding to full-length mouse Selenoprotein W protein. (Uniprot: P63300)
Molecular Weight of Antigen 9.54 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6415
Test Specificity This antibody is specific for mouse Selenoprotein W. A BLAST analysis was used to suggest cross-reactivity with SelW from rat based on a 98% identity with the immunizing sequence. Partial reactivity is expected against human-derived SelW based on an 87% homology with the immunogen. Cross-reactivity with SelW from other sources has not been determined.
Target Species Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.