Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Serotonin N-acetyltransferase, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB100135 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100135 0.1 mL
NB100135SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100135 Supplier Novus Biologicals Supplier No. NB100135

Rabbit Polyclonal Antibody has been used in 5 publications

Serotonin N-acetyltransferase Polyclonal specifically detects Serotonin N-acetyltransferase in Human, Primate, Bovine (Negative), Chicken (Negative), Rat (Negative), Zebrafish (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Serotonin N-acetyltransferase
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:200
Formulation Eluted in 0.1N glacial acetic acid containing 0.1% BSA and neutralized using 10x PBS with 0.05% Thimerosal
Gene Accession No. Q16613
Gene Alias AA-NAT, aralkylamine N-acetyltransferaseEC 2.3.1.87, arylalkylamine N-acetyltransferase, Serotonin acetylase, SNATserotonin N-acetyltransferase
Gene Symbols AANAT
Host Species Rabbit
Immunogen Human Serotonin N-acetyltransferase [UniProt# Q16613]
Molecular Weight of Antigen 23 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Circadian Rhythm, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 15
Test Specificity This is specific for primate AANAT (3,4).
Target Species Human, Primate, Bovine (Negative), Chicken (Negative), Rat (Negative), Zebrafish (Negative)
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.