Learn More
anti-Siglec-9, Fluorescein, Clone: 191240, R&D Systems™
Manufacturer:  R&D Systems FAB1139F100
 View more versions of this product
Catalog No. FAB1139F100
Add to cart
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
Siglec-9
191240
Flow Cytometry 10 uL/10^6 cells
Q9Y336
CD329, CDw329, FOAP-9, OBBP-Like, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9, Siglec9
Mouse
IgG2a
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human Siglec-9 in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Siglec-2, -3, -5, -7 or -10 is observed.
Flow Cytometry
Fluorescein
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
SIGLEC9
Mouse myeloma cell line NS0-derived recombinant human Siglec-9, Gln18-Gly348 (predicted), Accession # Q9Y336
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
27180
Human