Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SMAD1/5/8/9, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB10056443T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056443T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056443T Supplier Novus Biologicals Supplier No. NB10056443SS

Rabbit Polyclonal Antibody has been used in 1 publication

SMAD1/5/8/9 Polyclonal specifically detects SMAD1/5/8/9 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen SMAD1/5/8/9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/mL
Formulation PBS, 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q15797
Gene Alias BSP1, BSP-1, hSMAD1, JV4-1SMAD, mothers against DPP homolog 1 (Drosophila), MAD homolog 1, MADH1JV41, MADR1MAD, mothers against decapentaplegic homolog 1 (Drosophila), Mad-related protein 1, mothers against decapentaplegic homolog 1, Mothers against DPP homolog 1, SMAD family member 1SMAD 1, SMAD, mothers against DPP homolog 1, Smad1, TGF-beta signaling protein 1, transforming growth factor-beta signaling protein 1
Gene Symbols SMAD1
Host Species Rabbit
Immunogen This antibody was developed against two synthetic peptides corresponding to amino acids 18-32 (LGWKQGDEEEKWAEK) and amino acids 315-330 (SNVNRNSTIENTRRHI) of human SMAD1. GenBank Accession No. AAP36050.1.
Molecular Weight of Antigen 52 kDa
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 4086
Test Specificity In A549, a 50 kDa band is observed. The two synthetic peptide immunogens (LGWKQGDEEEKWAEK) and (SNVNRNSTIENTRRHI) used to make this antibody are 100% conserved between SMAD1/5/8/9. Therefore, the antibody recognizes SMADs 1, 5, 8, and 9. The antibody may cross-react with SMAD2 or SMAD3. The actual SMADs detected with this antibody may vary according to cell or tissue-type specific expression.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.