Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SOD1/Cu-Zn SOD, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB200600 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200600 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB200600 Supplier Novus Biologicals Supplier No. NB200600

Sheep Polyclonal Antibody

SOD1/Cu-Zn SOD Polyclonal antibody specifically detects SOD1/Cu-Zn SOD in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen SOD1/Cu-Zn SOD
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 10.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, ELISA 1:100-1:2000
Formulation Glycine Buffered Saline (pH 7.4), 0.01% Benzamidine, 0.1% EACA and 1.0mM EDTA with 0.09% Sodium Azide
Gene Alias ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic
Gene Symbols SOD1
Host Species Sheep
Immunogen Human SOD Cu,Zn purified from human erthrocytes.
Purification Method Immunogen affinity purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6647
Test Specificity Recognises superoxide dismutase.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.