Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPO11 meiotic protein covalently bound to DSB homolog (S. cerevisiae), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017035 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-035 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-035 Supplier Abnova Corporation Supplier No. H00023626B01P
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human SPO11 protein.

Meiotic recombination and chromosome segregation require the formation of double-strand breaks (DSBs) in paired chromosome homologs. During meiosis in yeast, a meiotic recombination protein is covalently-linked to the 5' end of DSBs and is essential for the formation of DSBs. The protein encoded by this gene is similar in sequence and conserved features to the yeast meiotic recombination protein. The encoded protein belongs to the TOP6A protein family. Several transcript variants encoding different isoforms have been found for this gene, but the full-length nature of only two of them have been described. [provided by RefSeq]

Sequence: MAFAPMGPEASFFDVLDRHRESLLAALRRGGREPPTGGSRLASRFEDSVGLQMVSHCTTRKIKSDSPKSAQKFSLILKILSMIYKLVQSNTYATKRDIYYTDSQLFGNQTVVDNIINDISCMLKVSRRSLHILSTSKGLIAGNLRYIEEDGTKVNCTCGATAVAVPSNIQGIRNLVTDAKFVLIVEKDATFQRLLDDNFCNKLSPCIMITGKGVPDLNTRLLVKKLWDTFHVPVFTLVDADPHGIEIMCIYKYGSMSMSFEAHHLTVPAIRWLGLLPSDLKRLNVPKDSLIPLTKRDQMKLDSILRRPYVTCQPFWRKEMEIMADSKMKAEIQALTFLSSDYLSRVYLPNKLKFGGWI

Specifications

Antigen SPO11 meiotic protein covalently bound to DSB homolog (S. cerevisiae)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human SPO11 protein.
Formulation PBS with no preservative; pH 7.4
Gene SPO11
Gene Accession No. NM_198265
Gene Alias MGC39953
Gene Symbols SPO11
Host Species Mouse
Immunogen SPO11 (NP_937998, 1 a.a. ∼ 358 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23626
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.