Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Substance P, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB300187 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300187 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300187 Supplier Novus Biologicals Supplier No. NB300187

Guinea Pig Polyclonal Antibody has been used in 7 publications

Substance P Polyclonal specifically detects Substance P in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Substance P
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. P06767
Gene Alias neurokinin 1, neurokinin 2, neurokinin A, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma), neuropeptide gamma, neuropeptide K, NK2, NKA, NKNAneurokinin alpha, NPK, PPT, protachykinin-1, substance K, substance P, TAC2neuromedin L, tachykinin 2, tachykinin, precursor 1, tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2
Gene Symbols TAC1
Host Species Guinea Pig
Immunogen A synthetic peptide made between amino acid residues 1-11 of rat Substance P [CRPKPQQFFGLM].
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6863
Test Specificity Other species have not been tested.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.