Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Synaptophysin, Clone: SY38, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB525820UG/DEL
Description
Reference:
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blotting
Specifications
| Synaptophysin | |
| Unconjugated | |
| Vesicular fraction of bovine brain. The SY38 epitope is to a pentapeptide repeat structure in the carboxy-terminal cytoplasmic tail of synaptophysin. | |
| RUO | |
| Primary | |
| Semi-purified |
| Monoclonal | |
| Mouse | |
| 20 μg | |
| Neuroscience | |
| Avian, Bovine, Fish, Human, Mouse, Rat | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction