Learn More
MilliporeSigma™ anti-Synaptophysin, Clone: SY38, ™ (Chemicon™)
Specifications
| Antigen | Synaptophysin |
|---|---|
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Semi-purified |
| Host Species | Mouse |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
50-175-583
|
MilliporeSigma™
MAB525820UG/DEL |
20 μg | N/A | N/A | N/A | ||||
|
50-175-584
|
MilliporeSigma™
MAB525850UG/DEL |
50 μg | N/A | N/A | N/A | ||||
Description
Reference:
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
| Synaptophysin | |
| Unconjugated | |
| Mouse | |
| Neuroscience | |
| IgG1 κ |
| Monoclonal | |
| Semi-purified | |
| RUO | |
| Avian, Bovine, Fish, Human, Mouse, Rat | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.