Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TBP like protein TLP, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP131133 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP131133 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP131133 Supplier Novus Biologicals Supplier No. NBP131133

Rabbit Polyclonal Antibody

TBP like protein TLP Polyclonal specifically detects TBP like protein TLP in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TBP like protein TLP
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias 21 kDa TBP-like protein, STUD21-kDA TBP-like protein, TATA box binding protein-related factor 2, TATA box-binding protein-like protein 1, TATA box-binding protein-related factor 2, TBP-like 1, TBP-like factor, TBP-like protein 1, TBP-related factor 2, TBP-related protein, TLFMGC:8389, TLP21, TLPMGC:9620, TRF2Second TBP of unique DNA protein, TRP
Gene Symbols TBPL1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human TLP. The exact sequence is proprietary.
Molecular Weight of Antigen 21 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9519
Test Specificity Tested in human. Mouse reactivity determined by Western blot. Predicted cross-reactivity based on sequence identity: Xenopus laevis (93%), Xenopus tropicalis (93%), Dog (100%), and Chimpanzee (100%). Not yet tested in any other species.
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.