Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-TCR beta chain, FITC, Clone: H57-597,
Armenian Hamster Monoclonal Antibody
$131.23 - $189.28
Specifications
| Antigen | TCR beta chain |
|---|---|
| Clone | H57-597 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | FITC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABF1454MI
|
MilliporeSigma™
MABF1454 |
25 μg |
Each for $189.28
|
|
|||||
|
MABF1455MI
|
MilliporeSigma™
MABF1455 |
100 μg |
Each for $131.23
|
|
|||||
Description
Specifically detects TCR beta chain Clone: H57-597 in Mouse samples, and it is validated for Flow Cytometry
The H57-597 antibody is specific for the beta chain of the mouse T cell Receptor (TCR). This cell surface protein combines with a second protein chain (alpha chain) to form the alpha-beta TCR that is expressed by NK1.1+ thymocytes, NKT cells, and the majority of peripheral T cells. A small number of T cells may express an alternative heteromer of gamma/delta protein chains, known as the g/d TCR. These receptors participate in a complex with CD3, and with the co-receptors CD4 or CD8, to recognize and respond to antigens bound to MHC molecules on antigen-presenting cells. Such interactions promote T cell receptor signaling (T cell activation) and can result in a number of cellular responses including proliferation, differentiation, production of cytokines or activation-induced cell death.The H57-597 antibody is used as a phenotypic marker for T cells expressing the alpha-beta TCR. It is also widely used to cross-link surface TCR and thereby mimic TCR-mediated cell activation or induction of apoptosis. The antibody does not cross-react with cells expressing the g/d TCR.Specifications
| TCR beta chain | |
| Flow Cytometry | |
| FITC | |
| Armenian Hamster | |
| Inflammation, Immunology | |
| Purified Armenian Hamster monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
| Ptcra | |
| IgG | |
| Purified by size-exclusion chromatography |
| H57-597 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| P0C6B2 | |
| Proprietary immunogen | |
| Primary | |
| 2°C to 8°C for one year from date of shipment protected from light; Do not freeze |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title