Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TetR Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$206.50 - $499.50
Specifications
| Antigen | TetR |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100, ELISA 1:100-1:2000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB600234SS
![]() |
Novus Biologicals
NB600234SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB600234
![]() |
Novus Biologicals
NB600234 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
TetR Polyclonal specifically detects TetR in Non-species specific samples. It is validated for Western Blot, ELISA.Specifications
| TetR | |
| Western Blot 1:100, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Transcription Factors and Regulators | |
| PBS and 30% Glycerol with 0.02% Sodium Azide | |
| pSC101 tetR, Tet Repressor, Tet Repressor Protein, TetR, tetracycline repressor protein | |
| A synthetic peptide made to an internal portion of Escherichia coli TetR protein (between residues 25-75) [UniProt P04483] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Non-species specific | |
| P04483 | |
| 4924774 | |
| IgG | |
| Affinity Purified | |
| Non-species specific. This antibody is used to monitor the regulation of gene expression in mammalian cells. The TetR antibody is not reactive to human or other mammalian proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title