Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat TNF-alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $2020.00
Specifications
Antigen | TNF-alpha |
---|---|
Clone | 45418 |
Dilution | Western Blot 1 ug/mL, Neutralization 10-40 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, Neutralization, ELISA |
Classification | Monoclonal |
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Rat samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
TNF-alpha | |
Western Blot 1 ug/mL, Neutralization 10-40 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Mouse | |
Rat | |
P16599 | |
7124 | |
E. coli-derived recombinant rat TNF-alpha Accession # P16599 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects rat TNF-alpha in ELISAs and Western blots. In ELISAs, this antibody shows less than 3% cross-reactivity with recombinant mouse (rm) TNF-alpha and less than 0.2% cross-reactivity with rhTNF-alpha , rpTNF-alpha , and rhTNF-beta . In Western blots, this antibody shows 100% cross-reactivity with rmTNF-alpha and no cross-reactivity with rfeTNF-alpha . |
45418 | |
Western Blot, Neutralization, ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
TNF | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title