Learn More
TOM70 Antibody, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody has been used in 4 publications
Supplier:  Novus Biologicals NB11058347SS
 View more versions of this product
Catalog No. NB11058347T
Only null left
Add to Cart
Description
Description
TOM70 Polyclonal specifically detects TOM70 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
TOM70
Polyclonal
Western Blot 1:500, Simple Western 1:50, Immunocytochemistry/Immunofluorescence 1:100
O94826
TOMM70
A synthetic peptide made to a region within residues 1-100 of the human TOM70 protein. [Swiss-Prot# O94826]
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
PBS with No Preservative
FLJ90470, KIAA0719, mitochondrial import receptor subunit TOM70, Mitochondrial precursor proteins import receptor, TOM70, Translocase of outer membrane 70 kDa subunit, translocase of outer mitochondrial membrane 70 (yeast) homolog A, translocase of outer mitochondrial membrane 70 homolog A (S. cerevisiae), translocase of outer mitochondrial membrane 70 homolog A (yeast)
Rabbit
67 kDa
0.025 mL
Apoptosis, Cellular Markers, Mitochondrial Markers, Signal Transduction
9868
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only