Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOM70 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 4 publications

$206.50 - $501.50

Specifications

Antigen TOM70
Dilution Western Blot 1:500, Simple Western 1:50, Immunocytochemistry/Immunofluorescence 1:100
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11058347T
SDP
View Documents
Novus Biologicals
NB11058347SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB11058347
SDP
View Documents
Novus Biologicals
NB11058347
0.1 mL
Each for $501.50
Only null left
Add to Cart
 
Description

Description

TOM70 Polyclonal specifically detects TOM70 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

TOM70
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
Rabbit
Apoptosis, Cellular Markers, Mitochondrial Markers, Signal Transduction
PBS with No Preservative
FLJ90470, KIAA0719, mitochondrial import receptor subunit TOM70, Mitochondrial precursor proteins import receptor, TOM70, Translocase of outer membrane 70 kDa subunit, translocase of outer mitochondrial membrane 70 (yeast) homolog A, translocase of outer mitochondrial membrane 70 homolog A (S. cerevisiae), translocase of outer mitochondrial membrane 70 homolog A (yeast)
TOMM70
IgG
Protein A purified
67 kDa
Western Blot 1:500, Simple Western 1:50, Immunocytochemistry/Immunofluorescence 1:100
Polyclonal
Purified
RUO
Human, Mouse, Rat
O94826
9868
A synthetic peptide made to a region within residues 1-100 of the human TOM70 protein. [Swiss-Prot# O94826]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.