Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521583
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA521583 100 μL
1 options

Catalog No. PIPA521583

Supplier: Invitrogen™ PA521583

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, Neuro2A, GL261, C8D30, NIH-3T3, BCL-1, Raw264.7, C2C12, PC-12, Rat2. Predicted reactivity: Mouse (100%), Rat (100%), Dog (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes an enzyme, consisting of two identical proteins, which catalyzes the isomerization of glyceraldehydes 3-phosphate and dihydroxy-acetone phosphate in glycolysis and gluconeogenesis. Mutations in this gene are associated with triosephosphate isomerase deficiency. Pseudogenes have been identified on chromosomes 1, 4, 6 and 7. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPI1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene Tpi1
Gene Accession No. P17751, P48500, P60174
Gene Alias AI255506; epididymis secretory protein Li 49; HEL-S-49; Methylglyoxal synthase; MGC88108; TIM; Tpi; TPI1; Tpi-1; TPID; triosephosphate isomerase; triose-phosphate isomerase; triosephosphate isomerase 1; triose-phosphate isomerase TPI1; unnamed protein product; YD9609.05C; YDR050C
Gene Symbols Tpi1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region within amino acids 187 and 249 of Triosephosphate isomerase (Uniprot ID#P60174).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21991, 24849, 7167
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.