Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIB3 Rabbit anti-Human, Polyclonal, Invitrogen™

Catalog No. PIPA515479
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA515479 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA515479 Supplier Thermo Scientific Supplier No. PA515479

Rabbit Polyclonal Antibody

NPK is a putative protein kinase that is induced by the transcription factor NF-kappaB. The encoded protein is a negative regulator of NF-kappaB and can also sensitize cells to TNF- and TRAIL-induced apoptosis. In addition, this protein can negatively regulate the cell survival serine-threonine kinase AKT1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIB3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene TRIB3
Gene Accession No. Q96RU7
Gene Alias Neuronal cell death inducible putative kinase; SKIP3; TRIB3
Gene Symbols TRIB3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 15-45 amino acids from the N-terminal region of human TRIB3
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57761
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.