Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl-Histone H3 (Lys36) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$153.22 - $615.02

Specifications

Antigen trimethyl Histone H3 (Lys36)
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ABE435SMI
View Documents
MilliporeSigma
ABE435S
20 μL
Each for $153.22
Only null left
Add to Cart
 
ABE435MI
View Documents
MilliporeSigma
ABE435
200 μL
Each for $615.02
Only null left
Add to Cart
 
Description

Description

Anti-trimethyl-Histone H3 (Lys36) Antibody is a rabbit polyclonal antibody for detection of Histone H3 trimethylated at lysine 36. Also known as Anti-H3K36me3, this purified Ab is specificity verified by dot blot (DB) and has been validated in western blot, immunocytochemistry, DB, Chimmunoprecipitation, ChIP-seq.
Specifications

Specifications

trimethyl Histone H3 (Lys36)
Polyclonal
Rabbit
NP_003484
KLH-conjugated linear peptide corresponding to human Histone H3 trimethylated at Lys36.
Affinity Purified
Immunocytochemistry, Western Blot
Unconjugated
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
Primary
2-8°C for 12 months
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.