Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Tubulin Mouse, Alexa Fluor 488, Clone: 5H1, BD

Catalog No. BDB558605 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558605 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558605 Supplier BD Biosciences Supplier No. 558605
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tubulin is a highly conserved protein with a molecular weight of ∽50 kD. The self-assembly of tubulin leads to microtubules, hollow cylinders that are one of the major components of the eukaryotic cytoskeleton. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton. There are two main classes of tubulin isoforms, α - and β -tubulin, which are usually products of separate genes. Microtubules are made from protofilaments, strings of alternating α - and β -tubulin spaced 4nm apart and pointing in the same direction. Tubulin can be posttranslationally modified in several ways, including phosphorylation, acetylation, glutamylation, and detyrosination. For example, microtubules that turn over slowly tend to be acetylated and detyrosinated.

TRUSTED_SUSTAINABILITY

Specifications

Antigen β-Tubulin
Applications In vitro Assay
Classification Monoclonal
Clone 5H1
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Chinese Hamster, Human, Indian Muntjac, Mouse, Rat, Syrian Hamster
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.