Learn More
Specifications
Specifications
| Antigen | Tyrosine Hydroxylase |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | 0.1mL in 10mM HEPES (pH 7.5), 150mM NaCl, 0.1mg/mL BSA and 50% glycerol. Adequate amount of material to conduct 10 mini Western Blots. |
| Host Species | Rabbit |
| Immunogen | SDS-denatured tyrosine hydroxylase purified from pheochromocytoma of Rat |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
