Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubc9 Mouse, Unlabeled, Clone: 50, BD
Mouse Monoclonal Antibody
$526.00
Specifications
Antigen | Ubc9 |
---|---|
Clone | 50 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Progression of the mammalian cell cycle is primarily regulated by phosphorylation/dephosphorylation and synthesis/degradation of many key proteins. Ubiquitin, a soluble protein of 76 amino acids, is enzymatically attached to an e-NH2-Lys in a target protein. Ubiquitination is a hallmark for rapid protein degradation of the target protein in the proteosome (a cytoplasmic complex of proteases). Human homologs of the yeast ubiquitin-conjugating enzymes (Ubc) have been reported, including Ubc9. Ubc9 is 158 amino acids with an apparent molecular weight of 18kDa. Although ubiquitously expressed, the highest levels of Ubc9 are found in testis and thymus. Ubc9 was localized to the synaptonemal complex in male mouse sex chromosomes. Furthermore, Ubc9 interacts with the recombination protein Rad51, thus suggesting an important role for Ubc9 during meiosis.Specifications
Ubc9 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2a | |
Affinity Purified |
50 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Chicken, Human, Mouse, Rat | |
Human Ubc9 aa. 26-156 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title