Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Ubiquitin, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB120837 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120837 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120837 Supplier Novus Biologicals Supplier No. NBP234617JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGF-23 Monoclonal specifically detects FGF-23 in Human samples. It is validated for ELISA.

Specifications

Antigen Ubiquitin
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Gene Accession No. Q12841
Host Species Rabbit
Immunogen Ubiquitin isolated from cow erythrocytes and conjugated to chicken gammaglobulins.
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6233
Test Specificity This antibody reacts with ubiquitin from human and several other mammalian species. It reacts with the periphery of senile plaques and neurofibrillary tangles in Alzheimer's disease. Cross-reacts with Human, Rat, Mouse and Cow. Not yet tested in other species.
Target Species Bovine, Human, Mouse, Rat
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.