Learn More
Uncoupling Protein 2 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Uncoupling Protein 2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in PBS, pH 7.4 with 0.1% BSA as stabilizer and 0.02% sodium merthiolate. |
| Gene Symbols | UCP2; UCPH; SLC25A8 |
| Host Species | Rabbit |
| Immunogen | 14 aa peptide sequence near the C-terminus of the mouse UCP2 (Fleury et al. 1997). The peptide has no significant homology with UCP1 or UCP3 (Boss et al. 1997). This peptide is available for blocking as catalog number AG788. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.