Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VCAM-1/CD106 Antibody, R&D Systems™

Catalog No. BBA6
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BBA6 1 mg
BBA5 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BBA6 Supplier R&D Systems Supplier No. BBA6
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 33 publications

VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Human samples. It is validated for Western Blot, Immunoprecipitation, Adhesion Blockade, Immunocytochemistry.

Specifications

Antigen VCAM-1/CD106
Classification Monoclonal
Clone BBIG-V1
Conjugate Unconjugated
Host Species Mouse
Immunogen Activated HUVEC human umbilical vein endothelial cells.
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.