Learn More
Description
Specifications
Specifications
| Antigen | Versican |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Immunoglobulin was purified by affinity chromatography and is presented in PBS with no preservative Anti-GST has not been adsorbed. |
| Gene Symbols | VCAN; GHAP; DKFZp686K06110; WGN; PG-M; WGN1; ERVR; CSPG2 |
| Host Species | Rabbit |
| Immunogen | GST fusion protein containing amino acids 535-598 of mouse versican. |
| Quantity | 50 μg |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.