Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-WIPF1/WIP, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB10068147 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10068147 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10068147 Supplier Novus Biologicals Supplier No. NB10068147

Goat Polyclonal Antibody

WIPF1/WIP Polyclonal specifically detects WIPF1/WIP in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen WIPF1/WIP
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.03 - 0.1 ug/ml, Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_003378.3
Gene Alias WAS/WASL interacting protein family, member 1, WAS/WASL-interacting protein family member 1, WASP-interacting protein, WASPIP, WIPMGC111041, WiskProtein PRPL-2, WiskPRPL-2
Gene Symbols WIPF1
Host Species Goat
Immunogen Peptide with sequence C-KLARNESRSGSNRRE corresponding to C-Terminus according to NP_001070737.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7456
Test Specificity Both variants represent identical proteins
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.