Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WNK lysine deficient protein kinase 3, Mouse, Clone: 4H5, Abnova™

Catalog No. 89014331 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-331 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-331 Supplier Abnova Corporation Supplier No. H00065267M01
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant WNK3.

Members of the 'with no lysine' (WNK) kinase family, such as WNK3, are serine-threonine protein kinases that lack the almost invariant catalytic lysine in subdomain II, which is important for binding ATP in the catalytic site. Instead, these kinases have a conserved lysine in subdomain I that is thought to provide this function (Holden et al., 2004 [PubMed 15194194]).[supplied by OMIM

Sequence: MATDSGDPASTEDSEKPDGISFENRVPQVAATLTVEARLKEKNSTFSASGETVERKRFFRKSVEMTEDDKVAESSPKDERIKAAMNIPRVDKLPSNVLRG

Specifications

Antigen WNK lysine deficient protein kinase 3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4H5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant WNK3.
Formulation PBS with no preservative; pH 7.4
Gene WNK3
Gene Accession No. NM_020922
Gene Alias KIAA1566/PRKWNK3
Gene Symbols WNK3
Host Species Mouse
Immunogen WNK3 (NP_065973, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Kinases
Primary or Secondary Primary
Gene ID (Entrez) 65267
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.