Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WNT4 Rabbit anti-Human, Polyclonal, Invitrogen™

Catalog No. PIPA513641
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA513641 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA513641 Supplier Thermo Scientific Supplier No. PA513641

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology.

The WNT family consists of secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. WNT4 is a protein which shows 98% amino acid identity to the Wnt4 protein of mouse and rat. It play a concerted role in both the control of female development and the prevention of testes formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT4
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene WNT4
Gene Accession No. P56705
Gene Alias Protein Wnt-4, WNT4
Gene Symbols WNT4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 242-269 amino acids from the C-terminal region of human WNT4
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54361
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.