Learn More
Yes Mouse, Unlabeled, Clone: 1, BD
Mouse Monoclonal Antibody
$286.00 - $568.50
Specifications
| Antigen | Yes |
|---|---|
| Clone | 1 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Yes is a member of the Src family of tyrosine kinases. The cellular gene was initially identified as a homologue of v-yes, the oncogene of avian sarcoma virus Y73. At least nine different Src family members have been identified. yes, fyn, and Src are widely expressed in a variety of cell types, while the remaining members are primarily expressed in hematopoietic cells. Members of this family have several common features: they have unique N-terminal domains, they attach to cellular membranes through a myristylated N-terminus and they have homologus SH2, SH3, and catalytic domains. The common expression patterns of Yes, fyn, and Src suggest some overlapping functions. All three of these kinases can physically associate with a number of cell-surface molecules which appear to increase their catalytic activity.
Specifications
| Yes | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 1 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Chicken, Human, Mouse, Rat | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.