Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ZDHHC6, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP182132 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP182132 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP182132 Supplier Novus Biologicals Supplier No. NBP182132

Rabbit Polyclonal Antibody

ZDHHC6 Polyclonal specifically detects ZDHHC6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ZDHHC6
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200-1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias DHHC-6, EC 2.3.1, EC 2.3.1.-, FLJ21952, Transmembrane protein H4, Zinc finger DHHC domain-containing protein 6, Zinc finger protein 376, zinc finger, DHHC domain containing 6, zinc finger, DHHC-type containing 6, ZNF376probable palmitoyltransferase ZDHHC6
Gene Symbols ZDHHC6
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:QLYHRLSFGWNTVKIDMSAARRDPLPIVPFGLAAF
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64429
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.