Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger protein 19, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89006468 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
89-006-468 50 μL
1 options

Catalog No. 89-006-468

Supplier: Abnova Corporation H00007567B01

Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human ZNF19 protein.

The protein encoded by this gene contains a zinc finger, a nucleic acid-binding domain present in many transcription factors. This gene is located in a region next to ZNF23, a gene also encoding a zinc finger protein, on chromosome 16. [provided by RefSeq

Sequence: MAAMPLKAQYQEMVTFEDVAVHFTKTEWTGLSPVQRALYRSVMLENFGNLTALGYPVPKPALISLLERGDMAWGLEAQDDPPAERTKNVCKDVETNIDSESTLIQGISEERDGMMSHGQLKSVPQRTDFPETRNVEKHQDIPTVKNIQGKVPRIPCARKPFICEECGKSFSYFSYYARHQRIHTGEKPFECSECGKAFNGNSSLIRHQRIHTGERPYHCEECGRAFNDNANLIRHQRIHSGDRPYYCTECGNSFTSSSEFVIHQRIHTGEKPYECNECGKAFVGNSPLLRHQKIHTGEKPYECNECGKSFGRTSHLSQHQRIHTGEKPYSCKVCGQAFNFHTKLTRHQRIHSEEKPFDCVDCGKAFSAQEQLKRHLRIHTQESSYVCDECGKALTSKRNLHQHQRIHTGEKPYECSKYEKAFGTSSQLGHLEHVYSGEKPVLDICRFGLPEFFTPFYW

Specifications

Antigen zinc finger protein 19
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human ZNF19 protein.
Formulation No additive
Gene ZNF19
Gene Accession No. BC047091.1
Gene Alias KOX12/MGC51021
Gene Symbols ZNF19
Host Species Mouse
Immunogen ZNF19 (AAH47091.1, 1 a.a. ∼ 458 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 7567
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.