Antimony(III) chloride, ACS, 99.0% min, Alfa Aesar™

Manufacturer: Alfa Aesar 36281A1

Quantity 1kg

 View more versions of this product

Catalog No. AA36281A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10025-91-9
    Physical Form Crystalline
    Assay Percent Range 99% min.
    Molecular Formula SbCl3
    Formula Weight 228.11
    MDL Number MFCD00011212
    Synonym Antimony trichloride
    Chemical Name or Material Antimony(III) chloride, For analysis ACS
    Grade ACS Grade

    Specifications

    Formula Weight 228.11
    Quantity 1kg
    Color White
    DOT Information DOT Class: 8, Packaging Group: II
    EINECS Number 233-047-2
    Physical Form Crystalline
    Odor Pungent
    Assay Percent Range 99% min.
    Molecular Formula SbCl3
    Appearance White crystalline
    CAS 10025-91-9
    Synonym Antimony trichloride
    MDL Number MFCD00011212
    Health Hazard 1 H314-H411
    Chemical Name or Material Antimony(III) chloride, For analysis ACS
    Grade ACS Grade

    H314-H411

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit