Learn More
Antimony(III) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Antimony(III) chloride is used as a chlorinating agent, as a fireproofing agent in textiles; in bronzing steel and as a mordant in dyeing as well as a caustic in medicine. It is also used as an apoptosis agent that is used to detect cholesterols and carotenoids. It is also employed to detect certain compounds, such as cholesterols and carotenoids. This agent seems to cause dehydration of cholesterol to 3,5-cholestadiene and 2,4-cholestadiene which produce the blue color seen during the reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 223.5°C |
| Melting Point | 73.4°C |
| Physical Form | Crystalline Aggregates |
| Quantity | 2 g |
| UN Number | UN1733 |
| Merck Index | 14,707 |
| Solubility Information | Soluble in alcohol and acetone and acids. |
| IUPAC Name | trichlorostibane |
| Formula Weight | 228.11 |
| Percent Purity | 99.999% |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314 Causes severe skin burns and eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ANTIMONY TRICHLORIDE, SOLID |
| EINECSNumber | 233-047-2 |
| RTECSNumber | CC4900000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.