Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Anza™ 14 SalI

Click to view available options
Quantity:
1500 units
8000 units
Description
Includes
Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity
• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity
Certifications
CoA

Specifications
Specifications
Content And Storage | 400 μL Anza 14 SalI 1 mL Anza Buffer (10X) 1 mL Anza Red Buffer (10X) Store at -20°C. |
Shipping Condition | Approved for shipment on Wet or Dry Ice |
Methylation Sensitivity | CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive |
Enzyme | Sal I |
Compatible Buffer | Anza™ 10X Red Buffer, Anza™ 10X Buffer |
Sensitive to Heat Inactivation | Yes |
Optimal Reaction Temperature | 37°C |
Type IIS RE | No |
Quantity | 8000 units |
Product Type | Restriction Enzyme Cloning System |
Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction